2-[(5-Bromo-3-methyl-2-pyridinyl)amino]-2-methyl-1-propanol

Supplier: Matrix Scientific

053104-500MG 053104-2.5G
101902-306EA 838.46 USD
101902-306 101902-304
2-[(5-Bromo-3-methyl-2-pyridinyl)amino]-2-methyl-1-propanol
2-[(5-Bromo-3-methyl-2-pyridinyl)amino]-2-methyl-1-propanol

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂O
MDL Number: MFCD13562577
CAS Number: 1220037-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR