2-Chloro-N-(2-hydroxy-1,1-dimethylethyl)nicotinamide

Supplier: Matrix Scientific

053103-500MG 053103-2.5G
101902-300EA 364.62 USD
101902-300 101902-302
2-Chloro-N-(2-hydroxy-1,1-dimethylethyl)nicotinamide
2-Chloro-N-(2-hydroxy-1,1-dimethylethyl)nicotinamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68 g/mol
MDL Number: MFCD12818875
CAS Number: 125335-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR