N-Cyclopentyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053062-2.5G 053062-500MG
101902-138EA 838.46 USD
101902-138 101902-136
N-Cyclopentyl-2-pyrrolidinecarboxamide hydrochloride
N-Cyclopentyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O
MDL Number: MFCD13562549
CAS Number: 1236266-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR