2-Chloro-N-cyclopentylnicotinamide

Supplier: Matrix Scientific

053051-500MG 053051-2.5G
101902-094EA 838.46 USD
101902-094 101902-092
2-Chloro-N-cyclopentylnicotinamide
2-Chloro-N-cyclopentylnicotinamide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O
MDL Number: MFCD00848989
CAS Number: 57841-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR