2-(Cyclopentylamino)nicotinic acid
Supplier: Matrix Scientific
053049-500MG
053049-2.5G
101902-086EA
838.46
USD
101902-086
101902-084
2-(Cyclopentylamino)nicotinic acid
2-(Cyclopentylamino)nicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ |
MDL Number:
MFCD03114311 CAS Number: 460363-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...