N-(3-Hydroxypropyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Supplier: Matrix Scientific
053040-500MG
053040-2.5G
101902-050EA
838.46
USD
101902-050
101902-048
N-(3-Hydroxypropyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-(3-Hydroxypropyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₇ClN₄O₂ |
MDL Number:
MFCD13562539 CAS Number: 1220016-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...