N-(3-Hydroxypropyl)-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053039-2.5G
053039-500MG
101902-046EA
838.46
USD
101902-046
101902-044
N-(3-Hydroxypropyl)-4-piperidinecarboxamide hydrochloride
N-(3-Hydroxypropyl)-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O₂ |
MDL Number:
MFCD13562538 CAS Number: 1219963-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...