2-Amino-N-(3-hydroxypropyl)-3-methylbutanamide hydrochloride
Supplier: Matrix Scientific
053033-500MG
053033-2.5G
101902-022EA
838.46
USD
101902-022
101902-020
2-Amino-N-(3-hydroxypropyl)-3-methylbutanamide hydrochloride
2-Amino-N-(3-hydroxypropyl)-3-methylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₉ClN₂O₂ |
MDL Number:
MFCD13562532 CAS Number: 1236256-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...