3-Amino-N-(3-hydroxypropyl)propanamide hydrochloride
Supplier: Matrix Scientific
053032-2.5G
053032-500MG
101902-018EA
838.46
USD
101902-018
101902-016
3-Amino-N-(3-hydroxypropyl)propanamide hydrochloride
3-Amino-N-(3-hydroxypropyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₅ClN₂O₂ |
MDL Number:
MFCD13562531 CAS Number: 1220034-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...