3-[(5-Bromo-2-pyridinyl)amino]-1-propanol
Supplier: Matrix Scientific
053027-2.5G
053027-500MG
101901-996EA
364.62
USD
101901-996
101901-998
3-[(5-Bromo-2-pyridinyl)amino]-1-propanol
3-[(5-Bromo-2-pyridinyl)amino]-1-propanol
Hazard: Irritant
Formula:
C₈H₁₁BrN₂O |
MDL Number:
MFCD11119174 CAS Number: 1036563-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...