2-[(3-Hydroxypropyl)amino]nicotinic acid

Supplier: Matrix Scientific

053023-500MG 053023-2.5G
101901-980EA 364.62 USD
101901-980 101901-982
2-[(3-Hydroxypropyl)amino]nicotinic acid
2-[(3-Hydroxypropyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₃
MDL Number: MFCD13562525
CAS Number: 1220037-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR