3-[(6-Chloro-2-pyrazinyl)amino]-1-propanol
Supplier: Matrix Scientific
053021-2.5G
053021-500MG
101901-974EA
838.46
USD
101901-974
101901-972
3-[(6-Chloro-2-pyrazinyl)amino]-1-propanol
3-[(6-Chloro-2-pyrazinyl)amino]-1-propanol
Hazard: Irritant
Formula:
C₇H₁₀ClN₃O |
MDL Number:
MFCD13562523 CAS Number: 1138220-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...