3-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-propanol

Supplier: Matrix Scientific

053018-500MG 053018-2.5G
101901-962EA 987.79 USD
101901-962 101901-960
3-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-propanol
3-[(5-Bromo-4-methyl-2-pyridinyl)amino]-1-propanol

Hazard: Irritant

Formula: C₉H₁₃BrN₂O
MDL Number: MFCD13562521
CAS Number: 1220019-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR