N-(2-Methoxyethyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053010-2.5G
053010-500MG
101901-930EA
838.46
USD
101901-930
101901-928
N-(2-Methoxyethyl)-2-pyrrolidinecarboxamide hydrochloride
N-(2-Methoxyethyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O₂ |
MDL Number:
MFCD13562515 CAS Number: 1236255-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...