N-(3-Hydroxybutyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Supplier: Matrix Scientific
052988-500MG
052988-2.5G
101901-842EA
838.46
USD
101901-842
101901-840
N-(3-Hydroxybutyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-(3-Hydroxybutyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₉ClN₄O₂ MW: 274.75 g/mol |
MDL Number:
MFCD13562503 CAS Number: 1220034-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...