N-(3-Hydroxybutyl)-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052985-500MG
052985-2.5G
101901-830EA
838.46
USD
101901-830
101901-828
N-(3-Hydroxybutyl)-2-piperidinecarboxamide hydrochloride
N-(3-Hydroxybutyl)-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O₂ |
MDL Number:
MFCD13562500 CAS Number: 1236262-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...