4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-2-butanol
Supplier: Matrix Scientific
052968-2.5G
052968-500MG
101901-760EA
364.62
USD
101901-760
101901-762
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-2-butanol
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-2-butanol
Hazard: Irritant
Formula:
C₁₀H₁₂ClF₃N₂O |
MDL Number:
MFCD13562483 CAS Number: 1220034-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...