2-Amino-N-isobutyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052957-500MG 052957-2.5G
101901-716EA 364.62 USD
101901-716 101901-718
2-Amino-N-isobutyl-3-phenylpropanamide hydrochloride
2-Amino-N-isobutyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂O
MDL Number: MFCD13562474
CAS Number: 1236254-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR