4-Bromo-2-nitrophenyl-2-(3-piperidinyl)ethylether hydrochloride
Supplier: Matrix Scientific
049773-2.5G
049773-500MG
101888-992EA
364.62
USD
101888-992
101888-994
4-Bromo-2-nitrophenyl-2-(3-piperidinyl)ethylether hydrochloride
4-Bromo-2-nitrophenyl-2-(3-piperidinyl)ethylether hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈BrClN₂O₃ MW: 365.66 g/mol |
MDL Number:
MFCD13560175 CAS Number: 1220017-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...