2-Chloro-4-(1-methyl-1-phenylethyl)phenyl2-(4-piperidinyl)ethyl ether hydrochloride

Supplier: Matrix Scientific

049732-500MG 049732-2.5G
101888-828EA 364.62 USD
101888-828 101888-830
2-Chloro-4-(1-methyl-1-phenylethyl)phenyl2-(4-piperidinyl)ethyl ether hydrochloride
2-Chloro-4-(1-methyl-1-phenylethyl)phenyl2-(4-piperidinyl)ethyl ether hydrochloride

Hazard: Irritant

Formula: C₂₂H₂₉Cl₂NO
MDL Number: MFCD13560134
CAS Number: 1220030-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR