3-(3-Bromophenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049510-2.5G 049510-500MG
101887-944EA 364.62 USD
101887-944 101887-946
3-(3-Bromophenoxy)azetidine hydrochloride
3-(3-Bromophenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClNO
MDL Number: MFCD26142755
CAS Number: 1707367-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR