3-(2,3-Dichlorophenoxy)azetidine

Supplier: Matrix Scientific

049487-500MG 049487-2.5G
101887-852EA 364.62 USD
101887-852 101887-854
3-(2,3-Dichlorophenoxy)azetidine
3-(2,3-Dichlorophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₉Cl₂NO
MDL Number: MFCD12828793
CAS Number: 1219948-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR