3-(Pentyloxy)azetidine

Supplier: Matrix Scientific

049458-2.5G 049458-500MG
101887-738EA 838.46 USD
101887-738 101887-736
3-(Pentyloxy)azetidine
3-(Pentyloxy)azetidine

Hazard: Irritant

Formula: C₈H₁₇NO
MDL Number: MFCD13559921
CAS Number: 1220021-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR