3-(4-Bromo-2-propylphenoxy)azetidine

Supplier: Matrix Scientific

049399-500MG 049399-2.5G
101887-500EA 364.62 USD
101887-500 101887-502
3-(4-Bromo-2-propylphenoxy)azetidine
3-(4-Bromo-2-propylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₂H₁₆BrNO
MDL Number: MFCD13559869
CAS Number: 1220027-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR