1-Benzoyl-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-Benzoylpiperidine-2-carboxylic acid

049372-500MG 049372-2.5G
101887-392EA 364.62 USD
101887-392 101887-394
1-Benzoyl-2-piperidinecarboxylic acid
1-Benzoyl-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
MDL Number: MFCD02211013
CAS Number: 78348-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR