Methyl (2S,4S)-4-(benzoyloxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049368-2.5G 049368-500MG
101887-378EA 838.46 USD
101887-378 101887-376
Methyl (2S,4S)-4-(benzoyloxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(benzoyloxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆ClNO₄
MDL Number: MFCD13559850
CAS Number: 1354487-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR