3-Azetidinyl cyclopentanecarboxylate

Supplier: Matrix Scientific

049339-500MG 049339-2.5G
101887-260EA 364.62 USD
101887-260 101887-262
3-Azetidinyl cyclopentanecarboxylate
3-Azetidinyl cyclopentanecarboxylate

Hazard: Irritant

Formula: C₉H₁₅NO₂
MDL Number: MFCD13559830
CAS Number: 1220021-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR