2-[(Cyclobutylcarbonyl)(methyl)amino]acetic acid
Supplier: Matrix Scientific
049335-2.5G
049335-500MG
101887-246EA
381.97
USD
101887-246
101887-244
2-[(Cyclobutylcarbonyl)(methyl)amino]acetic acid
2-[(Cyclobutylcarbonyl)(methyl)amino]acetic acid
Hazard: Irritant
Formula:
C₈H₁₃NO₃ |
MDL Number:
MFCD09044812 CAS Number: 926222-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...