1-(Cyclopropylcarbonyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049313-2.5G 049313-500MG
101887-158EA 838.46 USD
101887-158 101887-156
1-(Cyclopropylcarbonyl)-3-piperidinecarboxylic acid
1-(Cyclopropylcarbonyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MDL Number: MFCD09040653
CAS Number: 926198-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR