2-(2-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride

Supplier: Matrix Scientific

049304-500MG 049304-2.5G
101887-120EA 364.62 USD
101887-120 101887-122
2-(2-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride
2-(2-Piperidinyl)ethyl cyclopropanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₂
MDL Number: MFCD13559809
CAS Number: 1219960-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR