3-Azetidinyl cyclopropanecarboxylate

Supplier: Matrix Scientific

049299-2.5G 049299-500MG
101887-100EA 364.62 USD
101887-100 101887-102
3-Azetidinyl cyclopropanecarboxylate
3-Azetidinyl cyclopropanecarboxylate

Hazard: Irritant

Formula: C₇H₁₁NO₂
MDL Number: MFCD13559804
CAS Number: 1220027-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR