4-Piperidinyl cyclopropanecarboxylate hydrochloride

Supplier: Matrix Scientific

Synonyms: Piperidin-4-yl cyclopropanecarboxylate hydrochloride

049298-500MG 049298-2.5G
101887-096EA 364.62 USD
101887-096 101887-098
4-Piperidinyl cyclopropanecarboxylate hydrochloride
4-Piperidinyl cyclopropanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClNO₂
MW: 205.69 g/mol
MDL Number: MFCD13559803
CAS Number: 1219968-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR