2-Methyl-N-(3-methylbutanoyl)alanine

Supplier: Matrix Scientific

049294-500MG 049294-2.5G
101887-080EA 364.62 USD
101887-080 101887-082
2-Methyl-N-(3-methylbutanoyl)alanine
2-Methyl-N-(3-methylbutanoyl)alanine

Hazard: Irritant

Formula: C₉H₁₇NO₃
MDL Number: MFCD13559801
CAS Number: 1219980-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR