1-(3-Methylbutanoyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049293-2.5G 049293-500MG
101887-076EA 364.62 USD
101887-076 101887-078
1-(3-Methylbutanoyl)-3-piperidinecarboxylic acid
1-(3-Methylbutanoyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MDL Number: MFCD08442737
CAS Number: 926225-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR