Methyl (2S,4S)-4-[(3-methylbutanoyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049287-500MG 049287-2.5G
101887-052EA 364.62 USD
101887-052 101887-054
Methyl (2S,4S)-4-[(3-methylbutanoyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(3-methylbutanoyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₄
MDL Number: MFCD13559799
CAS Number: 1354488-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR