3-Piperidinylmethyl-3-methylbutanoate hydrochloride

Supplier: Matrix Scientific

049280-2.5G 049280-500MG
101887-026EA 838.46 USD
101887-026 101887-024
3-Piperidinylmethyl-3-methylbutanoate hydrochloride
3-Piperidinylmethyl-3-methylbutanoate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClNO₂
MDL Number: MFCD13559792
CAS Number: 1220037-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR