4-Piperidinyl-3-methylbutanoate hydrochloride

Supplier: Matrix Scientific

049278-500MG 049278-2.5G
101887-016EA 364.62 USD
101887-016 101887-018
4-Piperidinyl-3-methylbutanoate hydrochloride
4-Piperidinyl-3-methylbutanoate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MW: 221.73 g/mol
MDL Number: MFCD13559790
CAS Number: 1219967-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR