3-Piperidinyl-3-methylbutanoate hydrochloride

Supplier: Matrix Scientific

049277-2.5G 049277-500MG
101887-012EA 364.62 USD
101887-012 101887-014
3-Piperidinyl-3-methylbutanoate hydrochloride
3-Piperidinyl-3-methylbutanoate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD13559789
CAS Number: 1220037-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR