2-Methyl-N-(4-piperidinyl)propanamide hydrochloride

Supplier: Matrix Scientific

049267-500MG 049267-2.5G
101886-974EA 838.46 USD
101886-974 101886-972
2-Methyl-N-(4-piperidinyl)propanamide hydrochloride
2-Methyl-N-(4-piperidinyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD11809618
CAS Number: 1170578-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR