2-(4-Piperidinyl)ethyl-2-methylpropanoate hydrochloride

Supplier: Matrix Scientific

049261-500MG 049261-2.5G
101886-948EA 364.62 USD
101886-948 101886-950
2-(4-Piperidinyl)ethyl-2-methylpropanoate hydrochloride
2-(4-Piperidinyl)ethyl-2-methylpropanoate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClNO₂
MDL Number: MFCD13559782
CAS Number: 1219948-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR