N-Propionyl-β-alanine

Supplier: Matrix Scientific

049255-500MG
101886-924EA 364.62 USD
101886-924
N-Propionyl-β-alanine
N-Propionyl-beta-alanine

Hazard: Irritant

Formula: C₆H₁₁NO₃
MW: 145.16 g/mol
MDL Number: MFCD09049912
CAS Number: 126424-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR