N-Propionylalanine

Supplier: Matrix Scientific

049250-500MG 049250-2.5G
101886-906EA 517.88 USD
101886-906 101886-904
N-Propionylalanine
N-Propionylalanine

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD00778495
CAS Number: 56440-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR