3-Piperidinylmethyl propanoate hydrochloride

Supplier: Matrix Scientific

049241-500MG 049241-2.5G
101886-868EA 364.62 USD
101886-868 101886-870
3-Piperidinylmethyl propanoate hydrochloride
3-Piperidinylmethyl propanoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD13559768
CAS Number: 1220020-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR