2-[(4-Methoxybenzoyl)(methyl)amino]acetic acid

Supplier: Matrix Scientific

049236-500MG 049236-2.5G
101886-848EA 364.62 USD
101886-848 101886-850
2-[(4-Methoxybenzoyl)(methyl)amino]acetic acid
2-[(4-Methoxybenzoyl)(methyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD09047586
CAS Number: 133604-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR