4-Piperidinyl-4-methoxybenzoate hydrochloride

Supplier: Matrix Scientific

049220-500MG 049220-2.5G
101886-786EA 838.46 USD
101886-786 101886-784
4-Piperidinyl-4-methoxybenzoate hydrochloride
4-Piperidinyl-4-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MW: 271.75 g/mol
MDL Number: MFCD13559755
CAS Number: 1220021-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR