2-[(Tetrahydro-2H-pyran-4-ylcarbonyl)amino]acetic acid
Supplier: Matrix Scientific
049210-500MG
049210-2.5G
101886-746EA
838.46
USD
101886-746
101886-744
2-[(Tetrahydro-2H-pyran-4-ylcarbonyl)amino]acetic acid
2-[(Tetrahydro-2H-pyran-4-ylcarbonyl)amino]acetic acid
Hazard: Irritant
Formula:
C₈H₁₃NO₄ |
MDL Number:
MFCD11169983 CAS Number: 1042644-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...