3-Piperidinylmethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Supplier: Matrix Scientific

049201-500MG 049201-2.5G
101886-708EA 364.62 USD
101886-708 101886-710
3-Piperidinylmethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride
3-Piperidinylmethyl tetrahydro-2H-pyran-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₂ClNO₃
MDL Number: MFCD13559744
CAS Number: 1220020-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR