N-(2-Phenylacetyl)-β-alanine
Supplier: Matrix Scientific
049197-2.5G
049197-500MG
101886-692EA
364.62
USD
101886-692
101886-694
N-(2-Phenylacetyl)-β-alanine
N-(2-Phenylacetyl)-beta-alanine
Hazard: Irritant
Formula:
C₁₁H₁₃NO₃ MW: 207.23 g/mol |
MDL Number:
MFCD00458998 CAS Number: 55154-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...