N-(2,2,2-Trichloroacetyl)-β-alanine

Supplier: Matrix Scientific

049177-2.5G 049177-500MG
101886-614EA 838.46 USD
101886-614 101886-612
N-(2,2,2-Trichloroacetyl)-β-alanine
N-(2,2,2-Trichloroacetyl)-beta-alanine

Hazard: Irritant

Formula: C₅H₆Cl₃NO₃
MW: 234.47 g/mol
MDL Number: MFCD13559728
CAS Number: 101642-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR