2-[(2,2,2-Trichloroacetyl)amino]acetic acid
Supplier: Matrix Scientific
049169-2.5G
049169-500MG
101886-582EA
838.46
USD
101886-582
101886-580
2-[(2,2,2-Trichloroacetyl)amino]acetic acid
2-[(2,2,2-Trichloroacetyl)amino]acetic acid
Hazard: Irritant
Formula:
C₄H₄Cl₃NO₃ |
MDL Number:
MFCD13559720 CAS Number: 15166-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...